ENA321766155 Enamine

2-{[(tert-butoxy)carbonyl]amino}-2-(2-methoxyphenyl)acetic acid

  • MDL number MFCD06797221
  • Cas Number : 179417-69-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 146...148
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-79089

Safety Information

msds https://www.enaminestore.com/catalog/EN300-79089
Availability details for
Quantity
  • Availability
  • Add to Cart