ENA321766160 Enamine

1-[(phenylcarbamoyl)methyl]piperidine-3-carboxylic acid

  • MDL number MFCD11606337
  • Cas Number : 1082939-15-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-79094

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-79094
Availability details for
Quantity
  • Availability
  • Add to Cart