ENA321766165 Enamine

2-(1-methyl-1H-pyrazol-4-yl)ethan-1-ol

  • MDL number MFCD09864193
  • Cas Number : 176661-75-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-78052

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-78052
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart