ENA321766168 Enamine

2-(5,5-dimethyl-2,4-dioxoimidazolidin-1-yl)acetic acid

  • MDL number MFCD08457314
  • Cas Number : 64942-63-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-79115

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-79115
Availability details for
Quantity
  • Availability
  • Add to Cart