ENA321766169 Enamine

1-(4-fluorophenyl)-3-methoxypropan-1-one

  • MDL number MFCD12058710
  • Cas Number : 1157687-47-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 30-31
Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-79117

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-79117
Availability details for
Quantity
  • Availability
  • Add to Cart