ENA321766175 Enamine

3-methanesulfonylpropane-1-sulfonyl chloride

  • MDL number MFCD17167268
  • Cas Number : 72315-12-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 62...64
Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-79130

Safety Information

msds https://www.enaminestore.com/catalog/EN300-79130
Availability details for
Quantity
  • Availability
  • Add to Cart