ENA321766178 Enamine

N-butylprop-2-enamide

  • MDL number MFCD00080433
  • Cas Number : 2565-18-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-79134

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-79134
Availability details for
Quantity
  • Availability
  • Add to Cart