ENA321766183 Enamine

5,6-dimethylpyridazin-3-amine

  • MDL number MFCD16871484
  • Cas Number : 17551-35-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-78057

Safety Information

msds https://www.enaminestore.com/catalog/EN300-78057
Availability details for
Quantity
  • Availability
  • Add to Cart