ENA321766187 Enamine

3-[(4-bromothiophen-2-yl)methyl]-5,5-dimethylimidazolidine-2,4-dione

  • MDL number MFCD16095067
  • Cas Number : 1249029-18-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-79167

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-79167
Availability details for
Quantity
  • Availability
  • Add to Cart