ENA321766191 Enamine

N'-hydroxy-2-[4-(4-methylphenyl)-1,3-thiazol-2-yl]ethanimidamide

  • Cas Number : 329267-09-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-79179

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-79179
Availability details for
Quantity
  • Availability
  • Add to Cart