ENA321766195 Enamine

N-(1-carbamothioyl-1-methylethyl)benzamide

  • MDL number MFCD12792086
  • Cas Number : 582289-89-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 167-168
Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-79184

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-79184
Availability details for
Quantity
  • Availability
  • Add to Cart