ENA321766197 Enamine

5-(1-methyl-1H-pyrazol-4-yl)-1,3,4-oxadiazol-2-amine

  • MDL number MFCD11168102
  • Cas Number : 1152906-12-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 253...255
Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-79187

Safety Information

msds https://www.enaminestore.com/catalog/EN300-79187
Availability details for
Quantity
  • Availability
  • Add to Cart