ENA321766198 Enamine

1-(pyrrolidin-1-yl)butan-2-amine

  • MDL number MFCD06446856
  • Cas Number : 883547-04-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-79188

Safety Information

msds https://www.enaminestore.com/catalog/EN300-79188
Availability details for
Quantity
  • Availability
  • Add to Cart