ENA321766199 Enamine

1-(1-phenylethyl)piperidin-4-amine

  • MDL number MFCD09949683
  • Cas Number : 68304-97-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-79189

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-79189
Availability details for
Quantity
  • Availability
  • Add to Cart