ENA321766202 Enamine

ethyl 2-hydroxy-2-(oxolan-3-yl)acetate

  • MDL number MFCD16693951
  • Cas Number : 221546-88-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-79193

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-79193
Availability details for
Quantity
  • Availability
  • Add to Cart