ENA321766208 Enamine

4-amino-N-{3-[(dimethylamino)methyl]phenyl}-1,2,5-oxadiazole-3-carboxamide

  • MDL number MFCD17112492
  • Cas Number : 1311314-30-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-79204

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-79204
Availability details for
Quantity
  • Availability
  • Add to Cart