ENA321766209 Enamine

3-bromo-4-methoxy-N-methylbenzene-1-sulfonamide

  • MDL number MFCD02135253
  • Cas Number : 358665-54-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-79205

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-79205
Availability details for
Quantity
  • Availability
  • Add to Cart