ENA321766210 Enamine

ethyl 4-amino-3-hydroxybenzoate

  • MDL number MFCD01320538
  • Cas Number : 87081-52-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 90-94
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-79206

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-79206
Availability details for
Quantity
  • Availability
  • Add to Cart