ENA321766212 Enamine

[cyclopropyl(isothiocyanato)methyl]cyclopropane

  • MDL number MFCD14676314
  • Cas Number : 1247603-60-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-79210

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-79210
Availability details for
Quantity
  • Availability
  • Add to Cart