ENA321766214 Enamine

3-(4-chlorophenyl)thiomorpholine

  • MDL number MFCD11557769
  • Cas Number : 1082847-79-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-79212

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-79212
Availability details for
Quantity
  • Availability
  • Add to Cart