ENA321766215 Enamine

1-(3-methoxypropyl)-1H-pyrazol-3-amine

  • MDL number MFCD12768275
  • Cas Number : 1179235-39-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-79223

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-79223
Availability details for
Quantity
  • Availability
  • Add to Cart