ENA321766217 Enamine

5-chloro-3-methyl-1-phenyl-1H-pyrazole

  • MDL number MFCD00067853
  • Cas Number : 1131-17-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-79228

Safety Information

msds https://www.enaminestore.com/catalog/EN300-79228
Availability details for
Quantity
  • Availability
  • Add to Cart