ENA321766219 Enamine

2-(ethylsulfanyl)phenol

  • MDL number MFCD00010815
  • Cas Number : 29549-60-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-79236

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-79236
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart