ENA321766225 Enamine

6-cyclopropylpyridazin-3-amine

  • MDL number MFCD09836134
  • Cas Number : 1159814-07-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 134-135
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-79277

Safety Information

signalword Warning
pcodes P264,P270,P280,P301+P312,P305+P351+P338,P330,P337+P313,P501
hcodes Xi,Xn
MSDS https://www.enaminestore.com/catalog/EN300-79277
pictograms GHS07
hstatements H302,H319
Availability details for
Quantity
  • Availability
  • Add to Cart