ENA321766233 Enamine

4-(methanesulfonylmethyl)benzene-1-sulfonyl chloride

  • MDL number MFCD16666841
  • Cas Number : 1247407-55-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-79334

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-79334
Availability details for
Quantity
  • Availability
  • Add to Cart