ENA321766238 Enamine

(isocyanatomethyl)cyclopropane

  • MDL number MFCD16665719
  • Cas Number : 25694-89-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -20
Shipping Temperature Dry Ice
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-77870

Safety Information

signalword Danger
pcodes P210,P233,P240,P241,P242,P243,P260,P261,P264,P270,P271,P280,P284,P285,P301+P312,P301+P330+P331,P302+P352,P303+P361+P353,P304+P340,P304+P341,P305+P351+P338,P310,P312,P320,P330,P342+P311,P361,P363,P370+P378,P403+P233,P403+P235,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-77870
pictograms GHS02,GHS05,GHS06,GHS08
hstatements H225,H302,H311,H314,H330,H334
Availability details for
Quantity
  • Availability
  • Add to Cart