ENA321766239 Enamine

methyl 4,5-dimethyl-1H-indole-2-carboxylate

  • MDL number MFCD16065778
  • Cas Number : 1202240-82-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 144...146
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-79354

Safety Information

msds https://www.enaminestore.com/catalog/EN300-79354
Availability details for
Quantity
  • Availability
  • Add to Cart