ENA321766244 Enamine

methyl 3-(4-methoxyphenoxy)propanoate

  • MDL number MFCD10694158
  • Cas Number : 18333-12-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-79361

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-79361
Availability details for
Quantity
  • Availability
  • Add to Cart