ENA321766250 Enamine

1-[2-(difluoromethoxy)phenyl]ethan-1-ol

  • MDL number MFCD08442833
  • Cas Number : 886859-40-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-79368

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-79368
Availability details for
Quantity
  • Availability
  • Add to Cart