ENA321766251 Enamine

5-(4-fluoro-2-methylphenyl)furan-2-carbaldehyde

  • MDL number MFCD11208126
  • Cas Number : 1001010-58-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-78072

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-78072
Availability details for
Quantity
  • Availability
  • Add to Cart