ENA321766253 Enamine

7-tert-butylazepan-2-one

  • MDL number MFCD18917297
  • Cas Number : 155204-32-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 126...128
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-78074

Safety Information

msds https://www.enaminestore.com/catalog/EN300-78074
Availability details for
Quantity
  • Availability
  • Add to Cart