ENA321766255 Enamine

methyl 3-[(oxolan-2-ylmethyl)amino]propanoate

  • MDL number MFCD10687283
  • Cas Number : 1040688-74-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. -20
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-77871

Safety Information

msds https://www.enaminestore.com/catalog/EN300-77871
Availability details for
Quantity
  • Availability
  • Add to Cart