ENA321766256 Enamine

N-(1-carbamothioylpropyl)benzamide

  • MDL number MFCD12792661
  • Cas Number : 97723-93-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-78078

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-78078
Availability details for
Quantity
  • Availability
  • Add to Cart