ENA321766257 Enamine

N-(4-amino-2,6-dichlorophenyl)pyrazine-2-carboxamide

  • MDL number MFCD09043065
  • Cas Number : 926266-82-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-79398

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-79398
Availability details for
Quantity
  • Availability
  • Add to Cart