ENA321766259 Enamine

7-chloro-5-methyl-2,3-dihydro-1H-indole-2,3-dione

  • MDL number MFCD06260739
  • Cas Number : 155184-80-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-79404

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-79404
Availability details for
Quantity
  • Availability
  • Add to Cart