ENA321766263 Enamine

5-fluoro-2-[methyl(2-methylbutyl)amino]benzaldehyde

  • MDL number MFCD11639803
  • Cas Number : 1096809-56-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-79409

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-79409
Availability details for
Quantity
  • Availability
  • Add to Cart