ENA321766267 Enamine

7-chloro-2-methyl-2,3-dihydro-1-benzofuran

  • MDL number MFCD12197293
  • Cas Number : 57899-12-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-77872

Safety Information

msds https://www.enaminestore.com/catalog/EN300-77872
Availability details for
Quantity
  • Availability
  • Add to Cart