ENA321766270 Enamine

methyl 3-[(3-ethoxypropyl)amino]-2-methylpropanoate

  • MDL number MFCD12193104
  • Cas Number : 1154921-20-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-77873

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-77873
Availability details for
Quantity
  • Availability
  • Add to Cart