ENA321766276 Enamine

[2-(difluoromethoxy)-3-methoxyphenyl]methanol

  • MDL number MFCD08442940
  • Cas Number : 926188-34-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-79436

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-79436
Availability details for
Quantity
  • Availability
  • Add to Cart