ENA321766285 Enamine

5-{[(3-methoxyphenyl)methyl]amino}-2,3-dihydro-1H-1,3-benzodiazol-2-one

  • MDL number MFCD07611113
  • Cas Number : 884993-10-6
  • Molecular Weight : 269.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-79457

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-79457
Availability details for
Quantity
  • Availability
  • Add to Cart