ENA321766289 Enamine

1-[(4-bromophenyl)methyl]-2-methyl-1H-imidazole

  • MDL number MFCD14647733
  • Cas Number : 777865-57-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-78086

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-78086
Availability details for
Quantity
  • Availability
  • Add to Cart