ENA321766291 Enamine

5-cyano-2-methylbenzene-1-sulfonyl chloride

  • MDL number MFCD09741164
  • Cas Number : 372198-49-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-77884

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-77884
Availability details for
Quantity
  • Availability
  • Add to Cart