ENA321766293 Enamine

1-[(4-chlorophenyl)sulfanyl]butan-2-one

  • MDL number MFCD16167335
  • Cas Number : 51828-54-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 53-55
Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-78091

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-78091
Availability details for
Quantity
  • Availability
  • Add to Cart