ENA321766295 Enamine

1-(2-fluorophenyl)-2-methanesulfonylethan-1-one

  • MDL number MFCD16860246
  • Cas Number : 1249532-55-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-77755

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-77755
Availability details for
Quantity
  • Availability
  • Add to Cart