ENA321766304 Enamine

2-{4-[(tert-butoxy)carbonyl]piperazin-1-yl}-2-methylpropanoic acid

  • MDL number MFCD16990554
  • Cas Number : 1240619-73-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-77887

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-77887
Availability details for
Quantity
  • Availability
  • Add to Cart