ENA332671903 Enamine

2-(piperazin-1-ylmethyl)-4-(propan-2-yl)morpholine

  • MDL number MFCD19381847
  • Cas Number : 1088710-28-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-70966

Safety Information

msds https://www.enaminestore.com/catalog/EN300-70966
Availability details for
Quantity
  • Availability
  • Add to Cart