ENA332671910 Enamine

1-[2-(ethylcarbamoyl)-1,2,3,4-tetrahydroisoquinolin-5-yl]-5-methyl-1H-1,2,3-triazole-4-carboxylic acid

  • MDL number MFCD19381855
  • Cas Number : 1315367-22-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 200...202
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-74042

Safety Information

msds https://www.enaminestore.com/catalog/EN300-74042
Availability details for
Quantity
  • Availability
  • Add to Cart