ENA332671926 Enamine

indolizine-2-carbonitrile

  • MDL number MFCD09879893
  • Cas Number : 153274-67-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-78381

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-78381
Availability details for
Quantity
  • Availability
  • Add to Cart