ENA332671987 Enamine

1-benzyl-5-methylpiperidin-3-amine

  • MDL number MFCD19381931
  • Cas Number : 1315367-65-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-78912

Safety Information

msds https://www.enaminestore.com/catalog/EN300-78912
Availability details for
Quantity
  • Availability
  • Add to Cart