ENA332672029 Enamine

3,3-diethyl-2,3-dihydro-1H-indole

  • MDL number MFCD19381974
  • Cas Number : 130546-01-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-79347

Safety Information

msds https://www.enaminestore.com/catalog/EN300-79347
Availability details for
Quantity
  • Availability
  • Add to Cart